3-Chloro-4-fluoro-DL-phenylglycine

Supplier: APOLLO SCIENTIFIC
Warning

PC0139-1G PC0139-5G
76562-366EA 92 USD
76562-366 76560-726
3-Chloro-4-fluoro-DL-phenylglycine
3-Chloro-4-fluoro-DL-phenylglycine
Formula: C₈H₇ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD01631397
CAS Number: 261762-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR