5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR9176-1G OR9176-250MG
76563-122EA 157.9 USD
76563-122 76560-316
5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole
5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole
Formula: C₁₁H₁₂N₂O
Melting Pt: 120…122.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08271942
CAS Number: 864068-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR