2-Methoxy-3-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR8870-100G OR8870-25G OR8870-5G
76561-898EA 269.94 USD
76561-898 76559-968 76563-384
2-Methoxy-3-pyridineboronic acid
2-Methoxy-3-pyridineboronic acid
Formula: C₆H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03411572
CAS Number: 163105-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR