4,6-Dibromo-2-chloroanisole

Supplier: APOLLO SCIENTIFIC
Warning

OR55675-500MG OR55675-1G
76562-210EA 241.42 USD
76562-210 76561-184
4,6-Dibromo-2-chloroanisole
4,6-Dibromo-2-chloroanisole
Formula: C₇H₅Br₂ClO
Boiling Pt: 269…272 °C
Melting Pt: 75…76 °C
Density: 2.213 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD22543700
CAS Number: 174913-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR