2-Bromo-6-nitrobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR55196-1G OR55196-250MG OR55196-5G
76563-324EA 54 USD
76563-324 76560-560 76560-106
2-Bromo-6-nitrobenzyl bromide
2-Bromo-6-nitrobenzyl bromide
Formula: C₇H₅Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD11036286
CAS Number: 58579-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR