Ethyl hydrogen malonate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR54513-25G
OR54513-5G
76559-652EA
113.08
USD
76559-652
76560-490
Ethyl hydrogen malonate
Ethyl hydrogen malonate
Formula:
C₅H₈O₄ Boiling Pt: 208 °C Melting Pt: –13 °C Density: 1.19 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020490 CAS Number: 1071-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...