(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2R)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, (R)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

OR40210-1G OR40210-5G
76562-556EA 78.44 USD
76562-556 76562-776
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid
Formula: C₁₁H₁₇NO₅
Melting Pt: 119…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09953681
CAS Number: 1212176-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR