Pyrazine-2-carboximidamide hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR3402-10G OR3402-5G
76560-504EA 49.9 USD
76560-504 76562-772 76562-088
Pyrazine-2-carboximidamide hydrochloride 97%
Pyrazine-2-carboximidamide hydrochloride
Formula: C5H7ClN4
Storage Temperature: Ambient
MDL Number: MFCD05663360
CAS Number: 138588-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR