4,5,7-Trichloroquinoline
Supplier: APOLLO SCIENTIFIC
OR309454-1G
OR309454-250MG
76575-372EA
394.22
USD
76575-372
76575-370
4,5,7-Trichloroquinoline
4,5,7-Trichloroquinoline
Formula:
C₉H₄Cl₃N Storage Temperature: Ambient |
MDL Number:
MFCD07787625 CAS Number: 23834-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...