5-Bromo-2-nitrophenol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR30929-10G
OR30929-1G
OR30929-5G
76575-320EA
30.56
USD
76575-320
76575-316
76575-318
5-Bromo-2-nitrophenol 97%
5-Bromo-2-nitrophenol
Formula:
C₆H₄BrNO₃ Melting Pt: 40…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095027 CAS Number: 27684-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...