7-Acetyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(1H-Indazol-7-yl)ethan-1-one1-(1H-Indazol-7-yl)ethanone

OR30887-250MG OR30887-1G
76575-276EA 33.99 USD
76575-276 76575-274
7-Acetyl-1H-indazole
7-Acetyl-1H-indazole
Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11869765
CAS Number: 1159511-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR