7-(Aminomethyl)-2-methyl-2H-indazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(2-Methyl-2H-indazol-7-yl)methylamine
OR30883-500MG
OR30883-250MG
76575-266EA
275
USD
76575-266
76575-264
7-(Aminomethyl)-2-methyl-2H-indazole
7-(Aminomethyl)-2-methyl-2H-indazole
Formula:
C₉H₁₁N₃ MW: 161.204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11869761 CAS Number: 1144044-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...