7-(Aminomethyl)-1H-indazole

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (1H-Indazol-7-yl)methylamine

OR30870-500MG OR30870-250MG
76575-258EA 239.4 USD
76575-258 76575-256
7-(Aminomethyl)-1H-indazole
7-(Aminomethyl)-1H-indazole
Formula: C₈H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD10696780
CAS Number: 944904-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR