Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR307299-5G OR307299-250MG OR307299-1G
76574-966EA 877.08 USD
76574-966 76574-962 76574-964
Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate
Formula: C₆H₈BrN₃O₂
MW: 234.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28955031
CAS Number: 1352903-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR