Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR307287-5G OR307287-1G OR307287-250MG
76574-932EA 811.88 USD
76574-932 76574-930 76574-928
Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate
Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate
Formula: C₆H₆Br₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22372689
CAS Number: 1017802-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR