1-Methyl-3-nitro-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR307160-1G OR307160-5G OR307160-25G
76574-780EA 20.26 USD
76574-780 76574-782 76574-784
1-Methyl-3-nitro-1H-pyrazole
1-Methyl-3-nitro-1H-pyrazole
Formula: C₄H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03444368
CAS Number: 54210-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR