Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-Hydroxysulfosuccinimide sodium saltsulfo-N-HydroxysuccinimideHydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium saltN-Hydroxy sulfosuccinimide sodium saltSulfo NHSN-Hydroxysulphosuccinimide sodium saltHydroxy-2,5-dioxopyrrolidine-3-sulfonic acid sodium saltN-Hydroxy sulphosuccinimide sodium saltSulpho-N-Hydroxysuccinimide

OR307159-1G OR307159-5G OR307159-25G
76574-774EA 31 USD
76574-774 76574-778 76574-776
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 99%
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)
Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR