1-Bromo-4-pentylbenzene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane

OR3070-10G OR3070-100G OR3070-25G
76574-716EA 23.85 USD
76574-716 76574-720 76574-718
1-Bromo-4-pentylbenzene
1-Bromo-4-pentylbenzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 203…204 °C
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00061113
CAS Number: 51554-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR