5-Cyclopropylnicotinic acid

Supplier: APOLLO SCIENTIFIC

OR306638-1G OR306638-250MG
76574-654EA 619.34 USD
76574-654 76574-652
5-Cyclopropylnicotinic acid
5-Cyclopropylnicotinic acid
Formula: C₉H₉NO₂
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD06802408
CAS Number: 893738-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR