2-Methyl-2H-indazole-4-boronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

OR30589-5G OR30589-1G OR30589-250MG
76562-034EA 461.46 USD
76562-034 76562-964 76561-532
2-Methyl-2H-indazole-4-boronic acid pinacol ester
2-Methyl-2H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11109437
CAS Number: 885698-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR