3-Bromo-o-toluic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR3055-25G OR3055-100G
76574-460EA 23.44 USD
76574-460 76574-462
3-Bromo-o-toluic acid
3-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00270097
CAS Number: 76006-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR