6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
6-Bromo-1,3-dihydro-2H-imidazo[4,5-b]pyridin-2-one
OR303626-5G
OR303626-1G
OR303626-500MG
76574-134EA
179.4
USD
76574-134
76574-132
76574-130
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
Formula:
C₆H₄BrN₃O Melting Pt: >350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00763285 CAS Number: 148038-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...