2-(Aminomethyl)-2-methyl-1,3-dioxolane

Supplier: APOLLO SCIENTIFIC
Warning

OR303185-100MG OR303185-1G OR303185-250MG
76574-060EA 27.6 USD
76574-060 76574-064 76574-062
2-(Aminomethyl)-2-methyl-1,3-dioxolane
2-(Aminomethyl)-2-methyl-1,3-dioxolane
Formula: C₅H₁₁NO₂
Boiling Pt: 73 °C (31 mm Hg)
Melting Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD10698199
CAS Number: 3289-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR