2-Acetylbenzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Acetylbenzenecarbonitrile

OR301156-1G OR301156-5G
76573-780EA 39.74 USD
76573-780 76573-782
2-Acetylbenzonitrile 97%
2-Acetylbenzonitrile
Formula: C₉H₇NO
Boiling Pt: 148 °C/12mmHg
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00272571
CAS Number: 91054-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR