2-(4-Bromophenyl)ethanol

Supplier: APOLLO SCIENTIFIC
Warning

OR30084-5G OR30084-100G OR30084-25G
76573-616EA 28.52 USD
76573-616 76573-620 76573-618
2-(4-Bromophenyl)ethanol
2-(4-Bromophenyl)ethanol
Formula: C₈H₉BrO
Boiling Pt: 138 °C/9mmHg
Density: 1.436 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002897
CAS Number: 4654-39-1
EINECS: 225-093-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR