3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR300407-1G OR300407-250MG
76573-566EA 159.94 USD
76573-566 76573-564
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD09749982
CAS Number: 1000341-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR