1-(5-Bromo-2-chloropyridin-4-yl)ethan-1-one

Supplier: APOLLO SCIENTIFIC
Warning

OR300394-1G OR300394-250MG
76573-554EA 556.2 USD
76573-554 76573-552
1-(5-Bromo-2-chloropyridin-4-yl)ethan-1-one
1-(5-Bromo-2-chloropyridin-4-yl)ethan-1-one
Formula: C₇H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD11847348
CAS Number: 1245915-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR