tert-Butyl 5-formylindoline-1-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Boc-5-formylindoline

OR300270-1G OR300270-250MG
76573-436EA 275.04 USD
76573-436 76573-434
tert-Butyl 5-formylindoline-1-carboxylate
tert-Butyl 5-formylindoline-1-carboxylate
Formula: C₁₄H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD12408236
CAS Number: 879887-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR