2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR300260-250MG OR300260-100MG
76573-424EA 1141.92 USD
76573-424 76573-422
2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine
2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine
Formula: C₇H₉BrN₂S
Storage Temperature: Ambient
MDL Number: MFCD22571362
CAS Number: 1280704-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR