5-Bromo-3-cyclopropylpyrazin-2-amine

Supplier: APOLLO SCIENTIFIC
Warning

OR300237-250MG OR300237-1G
76573-358EA 206.78 USD
76573-358 76573-360
5-Bromo-3-cyclopropylpyrazin-2-amine
5-Bromo-3-cyclopropylpyrazin-2-amine
Formula: C₇H₈BrN₃
Storage Temperature: Ambient
MDL Number: MFCD31556420
CAS Number: 1845754-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR