Isochroman

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4-Dihydro-1H-2-benzopyran

OR28756-250G OR28756-50G
76572-640EA 103.9 USD
76572-640 76572-638
Isochroman
Isochroman
Formula: C₉H₁₀O
Boiling Pt: 89…90 °C/12mmHg
Density: 1.067 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006913
CAS Number: 493-05-0
EINECS: 207-774-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR