2-Mercapto-1-methylimidazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: ThiamazoleMethimazole2-Mercapto-N-methyl-1H-imidazole2-Mercapto-N-methylimidazole2-Mercapto-1-methyl-1H-imidazole1-Methyl-1H-imidazole-2(3H)-thione

OR28714-25G OR28714-100G
76572-614EA 19 USD
76572-614 76572-616
2-Mercapto-1-methylimidazole
2-Mercapto-1-methylimidazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 280 °C
Melting Pt: 144…147 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00179321
CAS Number: 60-56-0
EINECS: 200-482-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR