Disodium-2,7-naphthalenedisulfonate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,7-Naphthalenedisulphonic acid disodium salt, Sodium naphthalene-2,7-disulphonate, 2,7-Naphthalenedisulfonic acid disodium salt, Sodium naphthalene-2,7-disulfonate, Ebert-Merz alpha-acid disodium salt

OR2861-100G OR2861-500G OR2861-25G
76572-506EA 23.44 USD
76572-506 76572-508 76572-504
Disodium-2,7-naphthalenedisulfonate
Disodium-2,7-naphthalenedisulfonate
Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066365
CAS Number: 1655-35-2
EINECS: 216-733-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR