N-Hydroxysuccinimide 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR28434-100G OR28434-250G OR28434-1KG
76572-406EA 20.26 USD
76572-406 76572-408 76572-410
N-Hydroxysuccinimide 99%
N-Hydroxysuccinimide
Formula: C₄H₅NO₃
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS Number: 6066-82-6
EINECS: 228-001-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR