3-Pyridinecarboxamide oxime

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Pyridylamidoxime, Nicotinamide Oxime

OR28384-100G OR28384-5G OR28384-25G
76572-402EA 37.7 USD
76572-402 76572-398 76572-400
3-Pyridinecarboxamide oxime
3-Pyridinecarboxamide oxime
Formula: C₆H₇N₃O
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00265955
CAS Number: 1594-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR