2-Chloro-6-hydrazinopyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (6-Chloro-2-pyridyl)hydrazine

OR2830-5G OR2830-25G
76572-372EA 20.77 USD
76572-372 76572-374
2-Chloro-6-hydrazinopyridine 95%
2-Chloro-6-hydrazinopyridine
Formula: C₅H₆ClN₃
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00214724
CAS Number: 5193-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR