trans-4-Chloro-4'-methylchalcone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2E)-3-(4-Chlorophenyl)-1-(4-methylphenyl)prop-2-en-1-one, (2E)-3-(4-Chlorophenyl)-1-(4-methylphenyl)-1-oxoprop-2-ene

OR27882-250MG OR27882-1G
76572-304EA 54.44 USD
76572-304 76572-306
trans-4-Chloro-4'-methylchalcone
trans-4-Chloro-4'-methylchalcone
Formula: C₁₆H₁₃ClO
MW: 256.727 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00016346
CAS Number: 37620-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR