2,6-Dimethyl-3-hydroxypyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Dimethylpyridin-3-ol, 3-Hydroxy-2,6-lutidine

OR27677-1G OR27677-5G OR27677-10G
76572-242EA 28.52 USD
76572-242 76572-244 76572-246
2,6-Dimethyl-3-hydroxypyridine
2,6-Dimethyl-3-hydroxypyridine
Formula: C₇H₉NO
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00228692
CAS Number: 1122-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR