6-Amino-5-bromonicotinonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR2759-1G OR2759-5G
76572-224EA 89.64 USD
76572-224 76572-226
6-Amino-5-bromonicotinonitrile
6-Amino-5-bromonicotinonitrile
Formula: C₆H₄BrN₃
Melting Pt: 194…195 °C
Storage Temperature: Ambient
MDL Number: MFCD02102412
CAS Number: 477871-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR