5-Bromo-2-ethoxynicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR2747-5G OR2747-1G OR2747-10G
76572-130EA 231.24 USD
76572-130 76572-128 76572-132
5-Bromo-2-ethoxynicotinic acid
5-Bromo-2-ethoxynicotinic acid
Formula: C₈H₈BrNO₃
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD09836166
CAS Number: 393184-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR