4-Methoxy-3-nitrobenzaldehyde
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR26454-5G
OR26454-25G
OR26454-1G
76571-852EA
77.42
USD
76571-852
76571-854
76571-850
4-Methoxy-3-nitrobenzaldehyde
4-Methoxy-3-nitrobenzaldehyde
Formula:
C₈H₇NO₄ Melting Pt: 97…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126498 CAS Number: 31680-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...