2-Bromo-4-nitroimidazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-5-nitro-1H-imidazole

OR2576-5G OR2576-10G
76571-680EA 119.48 USD
76571-680 76571-676 76571-678
2-Bromo-4-nitroimidazole 97%
2-Bromo-4-nitroimidazole
Formula: C₃H₂BrN₃O₂
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD09025308
CAS Number: 65902-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR