2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan

Supplier: APOLLO SCIENTIFIC
Warning

OR2452-1G OR2452-250MG
76571-400EA 706.26 USD
76571-400 76571-398
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan
Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06797643
CAS Number: 87776-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR