5-Thiophen-2-ylisoxazole-3-carbaldehyde 90%

Supplier: APOLLO SCIENTIFIC
Warning

OR23368-250MG OR23368-1G
76571-086EA 199.66 USD
76571-086 76571-088
5-Thiophen-2-ylisoxazole-3-carbaldehyde 90%
5-Thiophen-2-ylisoxazole-3-carbaldehyde
Formula: C₈H₅NO₂S
Melting Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD03086160
CAS Number: 465514-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR