4-Imidazol-1-ylbenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(1-Imidazolyl)benzaldehyde, 1-(4-Formylphenyl)imidazole, 4-(1H-Imidazol-1-yl)benzaldehyde

OR23299-1G OR23299-5G
76571-052EA 20.26 USD
76571-052 76571-054
4-Imidazol-1-ylbenzaldehyde
4-Imidazol-1-ylbenzaldehyde
Formula: C₁₀H₈N₂O
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00209977
CAS Number: 10040-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR