5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR23279-1G OR23279-5G OR23279-10G
76571-020EA 124.28 USD
76571-020 76571-022 76571-024
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde
Formula: C₉H₇NOS₂
Melting Pt: 130…131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682090
CAS Number: 499770-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR