2,1,3-Benzoxadiazole-5-carbaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzo[c][1,2,5]oxadiazole-5-carbaldehyde

OR23134-250MG OR23134-1G
76570-932EA 44.82 USD
76570-932 76570-934
2,1,3-Benzoxadiazole-5-carbaldehyde 97%
2,1,3-Benzoxadiazole-5-carbaldehyde
Formula: C₇H₄N₂O₂
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD01200350
CAS Number: 32863-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR