2-Amino-N-cyclohexylbenzamide

Supplier: APOLLO SCIENTIFIC
Warning

OR2218-1G OR2218-250MG OR2218-5G
76570-570EA 55.02 USD
76570-570 76570-568 76570-572
2-Amino-N-cyclohexylbenzamide
2-Amino-N-cyclohexylbenzamide
Formula: C₁₃H₁₈N₂O
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00060339
CAS Number: 56814-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR