2-Aminoquinoline 96%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Quinolin-2-ylamine
OR22144-250MG
OR22144-1G
76570-560EA
19.48
USD
76570-560
76570-558
2-Aminoquinoline 96%
2-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 125…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038079 CAS Number: 580-22-3 EINECS: 209-458-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...