4-Hydroxy-3-nitrobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR22139-25G OR22139-100G
76570-554EA 20.26 USD
76570-554 76570-556
4-Hydroxy-3-nitrobenzoic acid 98%
4-Hydroxy-3-nitrobenzoic acid
Formula: C₇H₅NO₅
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007119
CAS Number: 616-82-0
EINECS: 210-494-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR